Fisherbrand™ Zipper Seal Sample Bags
Zipper-locking feature securely seals bags
Description
Zipper-locking feature securely seals bags
- Durable, transparent polyethylene
- Ideal for collecting, storing, and protecting laboratory specimens and samples easily and conveniently
- Clear, print-free surface lets user mark and label each bag separately
Not suggested for use with liquids
Specifications
Not Autoclavable | |
Polyethylene | |
2 mil | |
12 in. | |
9 in. |
Zip Seal | |
Non-sterile | |
12 x 9 in. (30 x 23cm) | |
30 cm | |
23 cm |