missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Histoplast Paraffin Wax
Three blends to accommodate all specimen types
$200.97 - $216.29
Catalog Number | Mfr. No. | Product Type | Melting Point | For Use With (Application) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Melting Point | For Use With (Application) | Price | Quantity | |||
22-900-700
|
N/A | Paraffin | 54° to 57°C | General use and all histology applications |
Case of 8 Each for $214.00
|
|
|||
22-900-701
|
Fisherbrand™
22900701 |
Paraffin | 54° to 57°C | For large and dense specimens |
Case of 8 Each for $200.97
|
|
|||
22-900-702
|
Fisherbrand™
22900702 |
Paraffin | 50° to 54°C | For improved ribboning with less compression |
Case of 8 Each for $216.29
|
|
|||
Description
When purity matters, only the finest will do.
- Formulated using raw materials of only the highest purity
- Is not sticky or brittle while sectioning
- Provides consistent tissue ribbons every time
Histoplast PE
For general use and all histology applications
Histoplast IM
For use on specimens requiring more thorough infiltration (large and dense specimens)
Histoplast LP
A lower melting-point formula for improved ribboning with less compression
Specifications
Paraffin | |
1kg |