missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ FiveEasy™ Plus FP20 pH/mV Meters
Compact, benchtop meter provides high quality pH/mV measurements with simple click of a button.
Supplier: Mettler Toledo™ 30266627
Includes: Meter and charger only. (Does not include electrode holder, buffers or probe.)
Description
- Compact, space saving design with ABS housing
- Large, well-structured 4.3 in. segmented LCD display for easy viewing
- Intuitive button arrangement and simple menu for easy operation
- RS232 or USB ports to export data directly either to a printer or a PC for further processing
- ATC and MTC temperature compensation
- Five point (max.) linear only calibration with four predefined buffer groups
- BNC, Cinch, NTC 30kiloOhm sensor inputs
- Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
- Ideal for wide range of laboratory applications
- pH Range: -2.00 to 16.00
- pH Resolution: 0.01/0.1
- pH Accuracy: ±:0.01
- mV Range: 2000 to 2000mv
- mV Resolution: 1mV
- mV Accuracy: ±1mV
- Temperature Range: -5° to +105°C (23° to 221°F)
- Temperature Resolution: 0.1°C
- Temperature Accuracy: 0.3°C
- Memory Size: 200 measurements, current calibration
- Power Supply: 100 to 240V, 50/60Hz, 12 VDC
- Dimensions: 22.7 x 14.7 x 7cm (9 x 5.7 x 2.7 in.)
- Weight: 0.63kg (1.4 lb.)
- FP20 Meter only: Suited for applications that require a specific sensor not included in a kit
Specifications
FiveEasy Plus FP20-Basic | |
RS232, USB | |
0.3°C | |
Applications that require a specific sensor not included in a kit | |
-2.00 to 16.00 | |
2000 to 2000 | |
-5°C to +105°C | |
0.1°C | |
200 measurements, current calibration | |
22.7 x 14.7 x 7 cm (9 x 5.7 x 2.7 in.) | |
1.4 lb. | |
FP20 Meter Only | |
50/60 Hz |
±0.01 | |
±1 | |
FP20 pH meter, charger, test certificate, pH measurement guide | |
Linear/Segmented | |
0.01, 0.1 | |
1mV | |
23°F to 221°F | |
LCD | |
BNC, Cinch, NTC 30kiloOhm sensors | |
0.63 kg | |
ATC and MTC | |
120/240 V |