missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ FiveGo™ F2 pH/mV Meters

Portable meter provides reliable and high quality pH/mV measurements with simple click of a button

Supplier:  Mettler Toledo™ 30266881

 View more versions of this product

Catalog No. 01-912-359

Explore available promotions

Includes: LE427 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack, carrying case

Description

Description

Different combinations of portable meters, sensors and accessories are available, which are ideally suited for specific applications or samples.

  • Robust, IP67 waterproof ABS housing
  • Large, well-structured 3.1 in. segmented LCD display for easy viewing
  • Intuitive button arrangement and simple menu for easy operation
  • Ergonomic design provide comfort to both big and small hands
  • Foldable foot stand provides optimal viewing while situated on a lab bench
  • Withstands wet and demanding environments
  • Includes compact carrying case
  • Three point calibration with four predefined buffer groups
  • ATC and MTC temperature compensation
  • Features Automatic and manual endpoint recording, Acoustic endpoint signal, Visual endpoint signal
  • BNC, Cinch sensor inputs
  • Ideal for mobile applications in the laboratory, at-line, or outdoors
Specifications:
  • pH Range: 0.00 to 14.00
  • pH Resolution: 0.01
  • pH Accuracy: ±0.01mV
  • mV Range: -1999 to 1999mV
  • mV Resolution: 1mV
  • mV Accuracy: ±1mV
  • Temperature Range: 0° to +100°C (32° to 212°F)
  • Temperature Resolution: 0.1°C
  • Temperature Accuracy: ±0.5°C
  • Memory Size: 200 measurements, current calibration
  • Operating Hours: >200 hrs.
  • Power Supply: 4 x 1.5V AAA LRO3 or 4 x 1.2V AAA NiMH HR03
  • Dimensions: 18.8 x 7.7 x 3.3cm (97.4 x 3 x 1.3 in.)
  • Weight: 0.26kg (0.5 lb.)
Applications:
  • F2-Meter only: Suitable for applications that require a specific sensor not included in a kit
  • F2 Food Kit: Suitable for solid and semi-solid samples such as meat, fruit, cheese, and more
  • F2 Field Kit: Suitable for wide range of general aqueous samples and for measurements on the go
  • F2 Standard Kit: Suitable for wide range of general aqueous samples including emulsions and suspensions
Specifications

Specifications

±1
LE427 IP67 sensor, electrode clip, wrist strap, installation QuickGuide, pH buffer starter pack, carrying case
0 to 14
1mV
0.1°
200 measurements, current calibration
18.8 x 7.7 x 3.3 cm (97.4 x 3 x 1.3 in.)
0.5 lb.
F2 Food Kit
±0.5°C
For solid and semi-solid samples such as meat, fruit, cheese, and more
0.01
3
LCD
BNC, Cinch
0.26 kg
ATC and MTC
SDS
Documents

Documents

Promotions

Promotions