Globe Scientific Flanged Plug Caps For 13mm Tubes
Provide leak-resistance in test tubes
Supplier:  Globe Scientific 118240B
 View more versions of this product
Catalog No. 22-171-668
Explore available promotions
Description
Description
  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in nine colors for sample identification purposes
Specifications
Specifications
2 flexible flanges
LDPE
0.51 in.
13 mm Tubes
Flange Plug Cap
Blue
13 mm
Product Suggestions
Product Suggestions