missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Globe Scientific Flanged Plug Caps For 13mm Tubes

Provide leak-resistance in test tubes

Supplier:  Globe Scientific 118240K

 View more versions of this product

Catalog No. 22-171-671

Explore available promotions

Description

Description

  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in nine colors for sample identification purposes

Specifications

Specifications

2 flexible flanges
LDPE
0.51 in.
13 mm Tubes
Flange Plug Cap
Black
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions