GSC Go Science Crazy Flask Brush
Designed specifically to clean flasks of varying sizes and shapes. The GSC International Flask Brush is sized to work with flasks of specific capacities and features a plastic handle shaped to reach flask curves.
Supplier: GSC Go Science Crazy 410420
Alert:
Intended for ages 13 and up.
Description
- Available for a variety of flask volumes
- Plastic handle is shaped with a curve to reach into flask curves
- Natural bristles for effective cleaning without damaging surfaces
Specifications
Natural Bristles | |
40 mm | |
330 mm |
Plastic | |
500mL flasks | |
90 mm |