Thermo Scientific™ Flat PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
$266.29 - $266.29
Specifications
Color | Natural |
---|---|
Certifications/Compliance | Certified DNA/RNase Free |
Material | Polypropylene |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
AB-0851
|
Thermo Scientific™
AB0851 |
PCR Plate Cap Strip |
Case of 200 Each for $266.29
|
|
|||||
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Flat PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certified free from human DNA, RNases, and DNases.
Specifications
Natural | |
Certified DNA/RNase Free | |
Polypropylene |