missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Flavanone, 98+%, Thermo Scientific Chemicals

$73.22 - $231.92

Chemical Identifiers

CAS 487-26-3
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00006841
InChI Key ZONYXWQDUYMKFB-UHFFFAOYNA-N
Synonym flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
PubChem CID 10251
ChEBI CHEBI:5070
IUPAC Name 2-phenyl-2,3-dihydrochromen-4-one
SMILES O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2258709
View Documents
Thermo Scientific Chemicals
B2258709
10 g
Each for $73.22
 
AAB2258718
View Documents
Thermo Scientific Chemicals
B2258718
50 g
Each for $231.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

487-26-3
224.26
ZONYXWQDUYMKFB-UHFFFAOYNA-N
10251
2-phenyl-2,3-dihydrochromen-4-one
C15H12O2
MFCD00006841
flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
CHEBI:5070
O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1

Specifications

487-26-3
C15H12O2
10 g
flavanone, 2-phenylchroman-4-one, 2,3-dihydroflavone, 2-phenyl-4-chromanone, 4-flavanone, 2,3-dihydro-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-2-phenyl, 2-phenyl-2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-2-phenyl-4h-benzopyran-4-one, 2-phenyl-3,4-dihydro-2h-1-benzopyran-4-one
O=C1CC(OC2=CC=CC=C12)C1=CC=CC=C1
224.26
CHEBI:5070
≥98%
75°C to 79°C
MFCD00006841
183227
ZONYXWQDUYMKFB-UHFFFAOYNA-N
2-phenyl-2,3-dihydrochromen-4-one
10251
224.26
Flavanone
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 207-654-8

RTECSNumber : LK6906500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only