Learn More
Flavin adenine dinucleotide disodium salt hydrate, 94% (dry wt.), water <10%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A14495ME
Description
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFlavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.
Solubility
Soluble in water (50 mg/ml).
Notes
Store at 4°C. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Flavin adenine dinucleotide disodium salt hydrate | |
(dry wt.), water <10% | |
MFCD00151217 | |
flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn | |
XLRHXNIVIZZOON-SJFBGTSINA-L | |
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate | |
131675332 | |
94% |
1891059-93-0 | |
C27H31N9Na2O15P2 | |
500 mg | |
Soluble in water (50mg/ml). | |
[Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1 | |
829.52 | |
829.52 |
Chemical Identifiers
1891059-93-0 | |
829.52 | |
XLRHXNIVIZZOON-SJFBGTSINA-L | |
131675332 | |
[Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1 |
C27H31N9Na2O15P2 | |
MFCD00151217 | |
flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn | |
disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate |
Safety and Handling
EINECSNumber : 282-733-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only