missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ampac™ Flexibles SealPAK Heavy-Duty Pouches

Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents

Supplier:  Ampac™ 50824

 View more versions of this product

Catalog No. 01-812-25G



Explore available promotions

Description

Description

Ideal for applications requiring long-term storage, sharper objects, or solutions of formaldehyde, acids, alkalies, greases, oils and organic solvents

  • Boilable and freezable pouches can sustain temperatures from -56 to 115°C (-70 to 240°F)
  • 4.5 mils thick
  • Transparent, airtight and leak-proof
  • Can be sterilized with gamma, infrared and e-beam
  • Sealed pouches can also be surface sterilized using VHP

Order Info

Also available in heat-sealable tubular roll stock that allows you to create custom-size pouches.

Specifications

Specifications

96 oz.
Polyester
4.50 mil
16 in.
12 in.
Transparent
360/Cs.
16 x 12 in. (40.6 x 30.5cm)
40.6 cm
30.5 cm
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions