missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Flt-3 Inhibitor, Thermo Scientific Chemicals

$496.73 - $496.73

Chemical Identifiers

CAS 301305-73-7
Molecular Formula C18H20N2O4S
Synonym 2-(3,4-Dimethoxybenzoylamino)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64484MA
View Documents
Thermo Scientific Chemicals
J64484MA
10 mg
Each for $496.73
 
Description

Description

A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
Specifications

Chemical Identifiers

301305-73-7
C18H20N2O4S
2-(3,4-Dimethoxybenzoylamino)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide

Specifications

10 mg
2°C to 8°C
301305-73-7
360.43g/mol
Light Sensitive
Powder
COC1=C(OC)C=C(C=C1)C(=O)NC1=C(C(N)=O)C2=C(CCCC2)S1
360.43
White
A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
C18H20N2O4S
2-(3,4-Dimethoxybenzoylamino)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide
Signal Transduction Reagent
FSPQCTGGIANIJZ-UHFFFAOYSA-N
2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only