missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Flumazenil, 98%, Thermo Scientific Chemicals

Benzodiazepine antagonist

$823.60 - $2103.75

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15H14FN3O3
Molecular Weight (g/mol) 303.293
MDL Number MFCD00242764
InChI Key OFBIFZUFASYYRE-UHFFFAOYSA-N
Synonym flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
PubChem CID 3373
ChEBI CHEBI:5103
IUPAC Name ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62537MC
View Documents
Thermo Scientific Chemicals
J62537MC
100 mg
Each for $823.60
 
AAJ62537ME
View Documents
Thermo Scientific Chemicals
J62537ME
500 mg
Each for $2,103.75
 
Description

Description

Benzodiazepine antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78755-81-4
303.293
OFBIFZUFASYYRE-UHFFFAOYSA-N
3373
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
C15H14FN3O3
MFCD00242764
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
CHEBI:5103
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C

Specifications

78755-81-4
C15H14FN3O3
100 mg
flumazenil, anexate, flumazepil, romazicon, lanexat, mazicon, flumazenilum, flumazenilo, flumazenilum latin, flumazenilo spanish
OFBIFZUFASYYRE-UHFFFAOYSA-N
ethyl 8-fluoro-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
3373
303.29
Powder
White
MFCD00242764
14,4135
Soluble in DMSO to 25mM
CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)F)C
303.293
CHEBI:5103
98%
Flumazenil
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NI2922170

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only