missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoral-P (=4-Amino-3-penten-2-one) 98.0+%, TCI America™

Supplier:  TCI America A53501G

 View more versions of this product

Catalog No. A53501G


Explore available promotions

Description

Description

Specifications

Specifications

Fluoral-P (=4-Amino-3-penten-2-one) [Fluorimetric reagent for aldehydes.]
41°C
214°C
MFCD00043715
acetylacetonamine, fluoral-p, 4-amino-3-penten-2-one, e-4-aminopent-3-en-2-one, 3-penten-2-one, 4-amino, 3e-4-aminopent-3-en-2-one, 4-aminopent-3-en-2-one, 4-amino-pent-3-en-2-one, e-4-amino-3-pentene-2-one, 2-acetyl-1-amino-1-methylethylene
CC(=CC(=O)C)N
99.133
CHEBI:51695
≥98.0% (GC,T)
1118-66-7
White-Yellow
C5H9NO
1 g
OSLAYKKXCYSJSF-ONEGZZNKSA-N
(E)-4-aminopent-3-en-2-one
5367854
99.13
Crystalline Powder

Chemical Identifiers

1118-66-7
99.133
OSLAYKKXCYSJSF-ONEGZZNKSA-N
5367854
(E)-4-aminopent-3-en-2-one
C5H9NO
MFCD00043715
acetylacetonamine, fluoral-p, 4-amino-3-penten-2-one, e-4-aminopent-3-en-2-one, 3-penten-2-one, 4-amino, 3e-4-aminopent-3-en-2-one, 4-aminopent-3-en-2-one, 4-amino-pent-3-en-2-one, e-4-amino-3-pentene-2-one, 2-acetyl-1-amino-1-methylethylene
CHEBI:51695
CC(=CC(=O)C)N
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions