missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Crystal/Powder at 20°C | |
86-73-7 | |
115°C | |
C13H10 | |
White | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
C1C2=CC=CC=C2C2=CC=CC=C12 | |
166.22 | |
CHEBI:28266 | |
≥95.0% (GC) |
Fluorene | |
295°C | |
500 g | |
MFCD00001111 | |
3077 | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
9H-fluorene | |
6853 | |
166.22 |
Chemical Identifiers
86-73-7 | |
166.22 | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
6853 | |
9H-fluorene |
C13H10 | |
MFCD00001111 | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
CHEBI:28266 | |
C1C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
EINECSNumber : (4)-0643
RTECSNumber : LL5670000
TSCA : Yes