Molecular Probes™ FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins
FluoReporter™ Biotin Quantitation Assay Kit, for biotinylated proteins
Supplier: Molecular Probes™ F30751
Description
The FluoReporter™ Biotin Quantitation Assay Kit provides a sensitive fluorometric assay for determining the number of biotin labels on nucleic acid samples (i.e., cDNA). The assay uses Biotective™ Green reagent, which contains a fluorescent dye that is quenched by ligands occupying the biotin binding sites. In the presence of biotin, the quencher dye ligands are displaced and fluorescence occurs. The fluorescence is proportional to the amount of added biotin. The results can be read with any fluorescence-based microplate reader capable of detecting FITC dye.Important Features of FluoReporter™ Biotin Quantitation Assay Kits:
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Biotin Quantitation Assay Kit | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Biotin |
1 Kit | |
Fluorescence | |
FluoReporter™ | |
Proteins |
For Research Use Only. Not for use in diagnostic procedures.