Learn More
Thermo Scientific Chemicals Fluorescein
Supplier: Thermo Scientific Chemicals 119240250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluorescein | |
2321-07-5 | |
MFCD00005050 | |
Solubility in water: insoluble. Other solubilities: soluble in dilute alkalies and dilute acids, soluble in boiling alcohol and ether, insoluble in benzene and chloroform | |
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O | |
332.31 | |
CHEBI:31624 | |
Pure | |
25 g |
314.0°C to 316.0°C | |
C20H12O5 | |
19, V,6, 456 | |
GNBHRKFJIUUOQI-UHFFFAOYSA-N | |
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one | |
16850 | |
332.3 | |
Glass bottle |
Chemical Identifiers
2321-07-5 | |
332.31 | |
GNBHRKFJIUUOQI-UHFFFAOYSA-N | |
CHEBI:31624 | |
C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O |
C20H12O5 | |
MFCD00005050 | |
16850 | |
3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement:
Wear eye protection/face protection.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 219-031-8
RTECSNumber : LM5075000
TSCA : TSCA
RUO – Research Use Only