missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein, Sodium Derivative, Sodium Salt, BAKER™, J.T. Baker™
A grade of chemicals for laboratory use
Supplier: Avantor M43007
Specifications
| Fluorescein, Sodium Derivative, Sodium Salt | |
| Wide Mouth Amber Glass Bottle | |
| MFCD00167039 | |
| C.I. 45350 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 | |
| 376.28 |
| 518-47-8 | |
| C20H10Na2O5 | |
| 500 g | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate | |
| 376.28 |
Chemical Identifiers
| 518-47-8 | |
| 376.28 | |
| NJDNXYGOVLYJHP-UHFFFAOYSA-L | |
| disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate |
| C20H10Na2O5 | |
| MFCD00167039 | |
| C.I. 45350 | |
| [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12 |