missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Fluorescent Protein Marker

Obtain accurate molecular weight protein standards. Skip gel staining steps and estimate protein molecular weight using UV.

Supplier:  MP Biomedicals™ 08W00009

Catalog No. ICN08W00009


Explore available promotions

Description

Description

  • Contains a total of 9 fluorescent proteins ranging from 14 kDa to 200 kDa.
  • It is designed as a molecular weight standard for the Quick Fluorescent Protein Buffer, pre-stained protein samples, and other fluorescent protein samples.
  • Increased protein band intensity allows for better electrophoresis monitoring while performing assays.
  • This product can also be used as a molecular weight marker for Western blot experiments, and the membrane-transferred fluorescent markers can be visualized on a UV or LED transilluminator.
Specifications

Specifications

UV Light, LED Transilluminator
Fluorescent Protein Marker
5 μL
Fluorescent
1 Vial
9
Yes
14 to 200 kDa
Liquid
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.