MP Biomedicals™ Ultra-Fast Running Buffer for SDS-PAGE
Ultra-Fast Running Buffer for SDS-PAGE in powder form is used in combination with traditional Tris-Glycine SDS-PAGE protein gels to achieve rapid electrophoresis.
Supplier:  MP Biomedicals™ 08W00012
Catalog No. ICN08W00012
Explore available promotions
Description
Description
Compared with traditional running buffers, Ultra-Fast Running Buffer significantly shortens electrophoresis times and improves experiment efficiency.
  • Each packet can prepare 500 mL of 1X buffer
  • Shortens SDS-PAGE electrophoresis times to as low as 20 minutes at 250 V
  • High-resolution band separation
  • Reduced buffer volumes
Specifications
Specifications
1X
10 Packets
Room Temperature
Powder
Videos
SDS
Documents
Documents
Promotions
Promotions
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.