missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Tris-Buffered Saline (TBS) Powder
Buffer salts to make TBS for membrane washing.
Supplier: MP Biomedicals™ 08W00020
Description
- For membrane washing
- 1 packet makes up 500 mL
Specifications
1X | |
Membrane | |
10 Packets |
Membrane Washing | |
Powder | |
Buffer |
Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.