missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 10ppm F- Premixed with Equal Volume of TISAB II, Certified, 10.0ppm ±0.1ppm, LabChem™
Supplier: LabChem LC146401
Specifications
Fluoride Standard | |
10ppm F- Premixed with Equal Volume of TISAB II | |
Poly Bottle | |
91.839,2.9,2.85,2.2,0.2 | |
Passes Test | |
NaF | |
Soluble in water | |
Liquid | |
sodium fluoride | |
41.99 |
10.0ppm ±0.1ppm | |
7681-49-4,7647-14-5,64-19-7,1310-73-2,125572-95-4,7732-18-5 | |
500 mL | |
Colorless | |
FNa | |
MFCD00003524 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
[F-].[Na+] | |
41.99 | |
Certified |
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
sodium fluoride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature