missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, 10ppm F- Premixed with Equal Volume of TISAB II, Certified, 10.0ppm ±0.1ppm, LabChem™

Supplier:  LabChem LC146402

 View more versions of this product

Catalog No. LC146402


Explore available promotions

Description

Description

Specifications

Specifications

Fluoride Standard
10ppm F- Premixed with Equal Volume of TISAB II
Poly Bottle
91.839,2.9,2.85,2.2,0.2
Passes Test
NaF
Soluble in water
Liquid
sodium fluoride
41.99
10.0ppm ±0.1ppm
7681-49-4,7647-14-5,64-19-7,1310-73-2,125572-95-4,7732-18-5
1 L
Colorless
FNa
MFCD00003524
PUZPDOWCWNUUKD-UHFFFAOYSA-M
[F-].[Na+]
41.99
Certified

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
[F-].[Na+]
FNa
MFCD00003524
sodium fluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions