missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, 1ppm F- Premixed with Equal Volume of TISAB II, Certified, 1.00ppm ±0.05ppm, LabChem™

Supplier:  LabChem LC146201

 View more versions of this product

Catalog No. LC146201


Explore available promotions

Description

Description

Specifications

Specifications

Fluoride Standard
1ppm F- Premixed with Equal Volume of TISAB II
Poly Bottle
91.849,2.9,2.85,2.2,0.2
FNa
MFCD00003524
[F-].[Na+]
41.99
CHEBI:28741
Certified
Soluble in water
1.00ppm ±0.05ppm
7681-49-4,7647-14-5,64-19-7,1310-73-2,125572-95-4
500 mL
Colorless
NaF
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
5235
41.99
Passes Test
Liquid

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
FNa
MFCD00003524
5235
sodium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions