missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 1ppm F- Premixed with Equal Volume of TISAB II, Certified, 1.00ppm ±0.05ppm, LabChem™
Supplier: LabChem LC146202
Specifications
Fluoride Standard | |
1ppm F- Premixed with Equal Volume of TISAB II | |
Poly Bottle | |
91.849,2.9,2.85,2.2,0.2 | |
FNa | |
MFCD00003524 | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
Certified | |
Soluble in water |
1.00ppm ±0.05ppm | |
7681-49-4,7647-14-5,64-19-7,1310-73-2,125572-95-4 | |
1 L | |
Colorless | |
NaF | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
41.99 | |
Passes Test | |
Liquid |
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
CHEBI:28741 | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
5235 | |
sodium fluoride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature