missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, 5ppm F- Premixed with Equal Volume of TISAB II, Certified, 5.00ppm ±0.10ppm, LabChem™
$117.97 - $380.53
Chemical Identifiers
CAS | 7681-49-4 |
---|---|
Molecular Formula | FNa |
Molecular Weight (g/mol) | 41.99 |
MDL Number | MFCD00003524 |
InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
PubChem CID | 5235 |
ChEBI | CHEBI:28741 |
IUPAC Name | sodium fluoride |
SMILES | [F-].[Na+] |
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
CHEBI:28741 | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
5235 | |
sodium fluoride |
Specifications
5.00ppm ±0.10ppm | |
Poly Bottle | |
91.844,2.9,2.85,2.2,0.2 | |
FNa | |
MFCD00003524 | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
Certified | |
Soluble in water | |
Fluoride Standard, 5ppm F- Premixed with Equal Volume of TISAB II |
7681-49-4,7647-14-5,64-19-7,1310-73-2,125572-95-4 | |
1 L | |
Colorless | |
NaF | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
41.99 | |
Passes Test | |
Liquid |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature