missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluoride Standard, Certified, 10.0ppm ±0.1ppm (1mL = 0.01mg F-), LabChem™
Supplier: LabChem LC145701
Specifications
Fluoride Standard | |
10ppm (1mL = 0.01mg F-) | |
Poly Bottle | |
99.998,0.002 | |
FNa | |
MFCD00003524 | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
Certified | |
Soluble in water |
10.0ppm ±0.1ppm (1mL = 0.01mg F-) | |
7681-49-4,7732-18-5 | |
500 mL | |
Colorless | |
NaF | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
41.99 | |
Passes Test | |
Liquid |
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
CHEBI:28741 | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
5235 | |
sodium fluoride |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature