missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluoride Standard, Certified, 1000ppm ±10ppm (1mL = 1mg F-), LabChem™

Supplier:  LabChem LC146001

 View more versions of this product

Catalog No. LC146001


Explore available promotions

Description

Description

Specifications

Specifications

Fluoride Standard
1000ppm (1mL = 1mg F-)
Poly Bottle
99.78,0.22
FNa
MFCD00003524
[F-].[Na+]
41.99
CHEBI:28741
Certified
Soluble in water
1000ppm ±10ppm (1mL = 1mg F-)
7681-49-4,7732-18-5
500 mL
Colorless
NaF
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium fluoride
5235
41.99
Passes Test
Liquid

Chemical Identifiers

7681-49-4
41.99
PUZPDOWCWNUUKD-UHFFFAOYSA-M
CHEBI:28741
[F-].[Na+]
FNa
MFCD00003524
5235
sodium fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions