missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluorometholone 98.0+%, TCI America™
Supplier: TCI America F04141G
Specifications
Fluorometholone | |
White-Yellow | |
MFCD00056461 | |
fluorometholone, oxylone, flumetholon, fluormetholone, fml liquifilm, fluor-op, cortilet, delmeson, trilcin, fml forte | |
CC1CC2C3CCC(C3(CC(C2(C4(C1=CC(=O)C=C4)C)F)O)C)(C(=O)C)O | |
376.468 | |
CHEBI:31625 | |
≥98.0% (HPLC) |
426-13-1 | |
C22H29FO4 | |
1 g | |
FAOZLTXFLGPHNG-KNAQIMQKSA-N | |
(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-6,10,13-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one | |
9878 | |
376.47 | |
Crystalline Powder |
Chemical Identifiers
426-13-1 | |
376.468 | |
FAOZLTXFLGPHNG-KNAQIMQKSA-N | |
9878 | |
(6S,8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11,17-dihydroxy-6,10,13-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one |
C22H29FO4 | |
MFCD00056461 | |
fluorometholone, oxylone, flumetholon, fluormetholone, fml liquifilm, fluor-op, cortilet, delmeson, trilcin, fml forte | |
CHEBI:31625 | |
CC1CC2C3CCC(C3(CC(C2(C4(C1=CC(=O)C=C4)C)F)O)C)(C(=O)C)O |
Safety and Handling
RTECSNumber : TU3834100
TSCA : No