missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluticasone propionate, Thermo Scientific Chemicals
$131.08 - $131.08
Chemical Identifiers
CAS | 80474-14-2 |
---|---|
Molecular Formula | C25H31F3O5S |
Molecular Weight (g/mol) | 500.57 |
InChI Key | WMWTYOKRWGGJOA-ZHLGSTKJSA-N |
PubChem CID | 91820603 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC462101000
|
Thermo Scientific Chemicals
462101000 |
100 mg |
Each for $131.08
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80474-14-2 | |
500.57 | |
91820603 |
C25H31F3O5S | |
WMWTYOKRWGGJOA-ZHLGSTKJSA-N |
Specifications
80474-14-2 | |
C25H31F3O5S | |
Solubility in water: .. Other solubilities: soluble in dioxane | |
500.57 | |
500.57 |
272°C to 273°C | |
100 mg | |
WMWTYOKRWGGJOA-ZHLGSTKJSA-N | |
91820603 | |
Fluticasone propionate |
RUO – Research Use Only