Puritan™ Foam Tipped Swabs
 View more versions of this product
Catalog No. 14-960-3J
Explore available promotions
Description
Description
Used for removing residues and particles; Applying adhesives or other solutions; General cleaning swab; Soft, onn-abrasive foam tip; Can be used with IPA, DI water, or dry; Lint and residue free; Good water absorption; Made in USA
Specifications
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2cm
1.49cm
1.5cm
0.18 in.
Non-sterile
Reticulated
0.1 in.
Critical environment industry, removing residues and particles, applying adhesives or other solutions, general cleaning
0.25cm
5.87 in.
0.625 in.
0.47cm
Medical
Polypropylene
Foam