Puritan™ Foam Tipped Swabs
Supplier:  Puritan™ 1804PF
 View more versions of this product
Catalog No. 19-817-280
Explore available promotions
Description
Description
Used for removing residues and particles; Applying adhesives or other solutions; General cleaning swab; Soft, onn-abrasive foam tip; Can be used with IPA, DI water, or dry; Lint and residue free; Good water absorption; Made in USA
Specifications
Specifications
CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
4 in.
Swab
10.16cm
9.8cm
1.5cm
0.156 in.
Non-sterile
Foam
0.90 in.
Removing residues and particles, general cleaning
0.22cm
3.87 in.
0.625 in.
0.39cm
Medical
Polypropylene