missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Puritan™ Foam Tipped Swabs

Supplier:  Puritan™ 2514061PFBT

 View more versions of this product

Catalog No. 22-029-629


Explore available promotions

Description

Description

Used for removing residues and particles; Applying adhesives or other solutions; General cleaning swab; Soft, onn-abrasive foam tip; Can be used with IPA, DI water, or dry; Lint and residue free; Good water absorption; Made in USA
Specifications

Specifications

CE, ISO 9001:2008, ISO 13485:2003, FDA GMP's
6 in.
Swab
15.2cm
15.2cm
Tapered
0.35cm
Medical
Acetal
0.098 in.
Specimen collection, diagnostic sampling, recovery of microorganisms and fluids
0.25cm
5.9 in.
0.547 in.
1.3cm
0.141 in.
Sterile
Foam
SDS
Documents

Documents

Promotions

Promotions