Foamtec UltraMOP™ Self-Wringing Mop Heads DFS Item
Clean room sponge mop head refill for Roll-O-Matic™
Supplier:  Foamtec FS62012
Catalog No. 19-304-749
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Tear-resistant, highly absorbent polyurethane sponge
  • With stainless steel bracket
  • Pin-hole-free foam for abrasion resistance
  • Available in Bump-Proof™ micro fiber over foam versions
  • For wet mopping procedures in sterile and controlled environments
Specifications
Specifications
10 in.
Autoclavable
Designed for wet mopping procedures in sterile and controlled environments
Non-sterile
Stainless steel bracket
Product Suggestions
Product Suggestions