missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Folic acid, ≥95%, Cell Culture Reagent Grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219466505
Specifications
59-30-3 | |
5 g | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
6037 |
C19H19N7O6 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
441.40 | |
CHEBI:27470 |
Chemical Identifiers
59-30-3 | |
441.40 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
CHEBI:27470 |
C19H19N7O6 | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
6037 |