missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Folic acid, ≥95%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210172525
Specifications
Folic acid | |
Yellow to Orange | |
C19H19N7O6 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N | |
441.404 | |
CHEBI:27470 | |
Powder |
59-30-3 | |
≥95% | |
25 g | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid | |
6037 | |
441.4 |
Chemical Identifiers
59-30-3 | |
441.404 | |
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid | |
CHEBI:27470 | |
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N |
C19H19N7O6 | |
OVBPIULPVIDEAO-LBPRGKRZSA-N | |
6037 | |
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid |