missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Folic acid, ≥95%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210172505

 View more versions of this product

Catalog No. ICN10172505


Explore available promotions

Description

Description

Specifications

Specifications

Folic acid
Yellow to Orange
C19H19N7O6
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
441.404
CHEBI:27470
Powder
59-30-3
≥95%
5 g
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
6037
441.4

Chemical Identifiers

59-30-3
441.404
folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
CHEBI:27470
C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N
C19H19N7O6
OVBPIULPVIDEAO-LBPRGKRZSA-N
6037
(2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
Videos
SDS
Documents

Documents

Promotions

Promotions