missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde, 37% by Weight (Histological), Fisher Chemical™

Stabilized with 10 to 15% (v/v) methanol

$541.45 - $1513.42

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
F75P-20
SDS View Documents Promotion Details
Fisher Chemical
BPF75P20
20 L PolyPac™
Each for $1,513.42
 
F75P1GAL
View Documents Promotion Details
Thermo Fisher Scientific
F75P1GAL
1 gal. Poly Bottle
Each for $541.45
Case of 4 Each for $2,165.80
 
Description

Description

Specifications

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O

Specifications

50-00-0,67-56-1,7732-18-5
Colorless
36.5 to 38.0 %
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.03
CHEBI:16842
36.5 to 38.0%
Stabilized with 10-15% (v/v) Methanol
Formaldehyde, 37% by Weight
0°C
101°C
PolyPac™
HCHO
20 L
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
30.03
Histological
Liquid
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'