missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formalin, 10%, Neutral Buffered, Reagents
Formalin, 10%, Neutral Buffered, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C22085104A
Specifications
| Formalin, 10%, Neutral Buffered | |
| 100°C | |
| 100°C | |
| HCHO/H2O | |
| 1.02 | |
| Reagent |
| 7732-18-5,50-00-0,67-56-1,7558-79-4,10049-21-5 | |
| Colorless | |
| Poly Bottle | |
| 4 L | |
| Formalin | |
| Liquid |