missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formalin, Buffered, 10% (Phosphate Buffer/Certified), Fisher Chemical™ Edge
Edge

Supplier:  Thermo Fisher Scientific SF1004

 View more versions of this product

Catalog No. SF100-4



Edge
Explore available promotions

Description

Description

Specifications

Specifications

Liquid
0°C
93.9°C
1.10g/cm³
6.9 to 7.1
Specially filtered for use in tissue processors
MFCD00003274
Certified
Formalin, Buffered, 10%
Phosphate Buffer
50-00-0,67-56-1,7558-79-4,10049-21-5,7732-18-5
Poly Bottle
4 L
Assay: 4 to 5% w/w
4 to 5%
10% w/v (Phosphate Buffer)
SDS
Documents

Documents

Promotions

Promotions