missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Formamide, Deionized, OmniPur™, Calbiochem™,

Supplier:  MilliporeSigma™ 4650500ML

 View more versions of this product

Catalog No. 46-505-00ML


Explore available promotions

Description

Description

Specifications

Specifications

Formamide
75-12-7
1.13g/cm3 at 20°C
500 mL
Odorless
CH3NO
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
NC=O
45.04
CHEBI:48431
≥99%
2.9°C
Colorless
209.9°C
154.9°C (310.8°F), Open Cup
Liquid
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45
Molecular Biology

Chemical Identifiers

75-12-7
45.04
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
NC=O
Videos
Safety and Handling

Safety and Handling

H360D May damage the unborn child.

H319: Causes serious eye irritation.

Recommended Storage : 2° to 8°C

SDS
Documents

Documents

Promotions

Promotions