missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Formamide (Molecular Biology), Fisher BioReagents
For nucleic acid hybridization and sequencing
Supplier: Fisher BioReagents BP227500
Description
Molecular Biology-grade Formamide is commonly used in nucleic acid hybridization and sequencing. It requires pretreatment with a mixed-bed resin.Specifications
4 to 5 | |
2°C | |
0.1 max. at 280nm | |
210°C | |
Pass Test | |
Liquid | |
CH3NO | |
MFCD00007941 | |
3.75 mPa/s at 20°C | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
NC=O | |
45.04 | |
CHEBI:48431 | |
≥99% |
Formamide | |
75-12-7 | |
1.133g/cm³ | |
500 mL | |
Glass Bottle | |
≥99 % | |
H2NCH:O | |
0.08mbar at 20°C | |
15, 4265 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
formamide | |
713 | |
45.04 | |
Molecular Biology |
Chemical Identifiers
75-12-7 | |
45.04 | |
ZHNUHDYFZUAESO-UHFFFAOYSA-N | |
713 | |
formamide |
CH3NO | |
MFCD00007941 | |
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88 | |
CHEBI:48431 | |
NC=O |
Safety and Handling

Recommended Storage : 4°C