missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Formamide, Molecular Biology Grade Calbiochem™,

Adenine deaminase inhibitor

$56.91 - $372.00

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
344206100ML
View Documents
MilliporeSigma™
344206100ML
100 mL
Each for $56.91
 
3442061L
View Documents
MilliporeSigma™
3442061L
1 L
Each for $372.00
 
Description

Description

Specifications

Chemical Identifiers

75-12-7
45.04
ZHNUHDYFZUAESO-UHFFFAOYSA-N
713
formamide
CH3NO
MFCD00007941
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
CHEBI:48431
NC=O

Specifications

2.9°C
Colorless
100 mL
Odorless
CH3NO
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
NC=O
45.04
CHEBI:48431
Molecular Biology
75-12-7
209.9°C
154.9°C (310.8°F), Open Cup
Liquid
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
formamide
713
45
Formamide
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area.

SDS
missing translation for 'documents'

missing translation for 'documents'