Learn More
Formamidine disulfide dihydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2033106
Description
Formamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFormamidine disulfide dihydrochloride is used as an intermediate in agriculture and dyestuff intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications
Formamidine disulfide dihydrochloride | |
∼175°C (decomposition) | |
MFCD00012586 | |
formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride | |
BFJQSCVWXZOXGK-UHFFFAOYSA-N | |
carbamimidoylsulfanyl carbamimidothioate;dihydrochloride | |
24802135 | |
97% |
14807-75-1 | |
C2H8Cl2N4S2 | |
5 g | |
Soluble in water. | |
[Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N | |
223.13 | |
223.15 |
Chemical Identifiers
14807-75-1 | |
223.13 | |
BFJQSCVWXZOXGK-UHFFFAOYSA-N | |
24802135 | |
[Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N |
C2H8Cl2N4S2 | |
MFCD00012586 | |
formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride | |
carbamimidoylsulfanyl carbamimidothioate;dihydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-876-3
RTECSNumber : LQ4570000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only