missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formamidine hydrochloride, 98%, Thermo Scientific Chemicals

$95.36 - $341.33

Chemical Identifiers

CAS 6313-33-3
Molecular Formula CH4ClN2
Molecular Weight (g/mol) 79.51
MDL Number MFCD00012865
InChI Key OEJWQRAWUVWBIE-UHFFFAOYSA-N
Synonym formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
PubChem CID 10313058
IUPAC Name methanimidamide;hydrochloride
SMILES [Cl].NC=N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6094706
View Documents
Thermo Scientific Chemicals
H6094706
5 g
Each for $95.36
 
AAH6094714
View Documents
Thermo Scientific Chemicals
H6094714
25 g
Each for $341.33
 
Description

Description

5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-4,6-dihydroxypyrimidine was synthesized from formamidine hydrochloride. N-Formimino-n-glutamate was synthesized from formamidine hydrochloride. Formamidine hydrochloride was used in the synthesis of imidazoleglycerol phosphate (IGP).

Solubility
Soluble in Ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
Specifications

Chemical Identifiers

6313-33-3
79.51
OEJWQRAWUVWBIE-UHFFFAOYSA-N
10313058
[Cl].NC=N
CH4ClN2
MFCD00012865
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
methanimidamide;hydrochloride

Specifications

6313-33-3
CH4ClN2
MFCD00012865
3906935
formamidine hydrochloride, formimidamide hydrochloride, formamidinium chloride, methanimidamide, monohydrochloride, formamidine hcl, formimidamide hcl, formamidine hcl salt, imidoformamide hydrochloride, methanimidamide hydrochloride
OEJWQRAWUVWBIE-UHFFFAOYSA-N
methanimidamide;hydrochloride
10313058
98%
84°C to 87°C
HN=CHNH2·HCl
5 g
Hygroscopic
Soluble in Ethanol.
[Cl].NC=N
79.51
80.52
Formamidine hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-639-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only