Learn More


Formic Acid, 88% (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific BPA118P100
Specifications
Formic Acid, 88% | |
8°C | |
101°C | |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
Certified ACS | |
100 mL | |
Poly Bottle | |
44mbar at 20°C |
2.1 | |
64-18-6 | |
Colorless | |
HCOOH | |
1779 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.03 | |
1.220g/cm³ | |
50°C | |
Liquid |
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Safety and Handling


DANGER!
Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 8, : Corrosive
Recommended Storage : Store separately from sulfuric acid and caustics.