missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic Acid, 88% (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific BPA118P500

 View more versions of this product

Catalog No. A118P-500



Explore available promotions

Description

Description

Specifications

Specifications

Formic Acid, 88%
8°C
101°C
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
Certified ACS
500 mL
Liquid
2.1
64-18-6
Colorless
HCOOH
1779
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.03
1.220g/cm³
Poly Bottle
44mbar at 20°C

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 8, : Corrosive

Recommended Storage : Store separately from sulfuric acid and caustics.

SDS Documents

Documents

Accessories
Promotions

Promotions