missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formic acid, 98+%, pure, Thermo Scientific Chemicals
$69.97 - $459.31
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
Molecular Weight (g/mol) | 46.025 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
SMILES | C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147930250
|
Thermo Scientific Chemicals
147930250 |
25 mL | Plastic bottle |
Each for $69.97
|
|
||||
AC147932500
|
Thermo Scientific Chemicals
147932500 |
250 mL | Plastic bottle |
Each for $109.55
|
|
||||
AC147930010
|
Thermo Scientific Chemicals
147930010 |
1 L | Plastic bottle |
Each for $199.42
Case of 4 Each for $0.00
|
|
||||
AC147930025
|
Thermo Scientific Chemicals
147930025 |
2.5 L | Plastic bottle |
Each for $354.14
|
|
||||
AC147930100
|
Thermo Scientific Chemicals
147930100 |
10 L | Plastic drum |
Each for $459.31
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-18-6 | |
46.025 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
CH2O2 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
Specifications
64-18-6 | |
CH2O2 | |
02, 8 | |
15, 4269 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
Pure | |
25 mL | |
Plastic bottle | |
1.3701 to 1.3721 (20°C, 589nm) | |
Formic acid |
98+% | |
HCO2H | |
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥98.0% | |
1.2200g/mL | |
Authentic | |
Liquid | |
44mbar at 20°C |
Safety and Handling
Danger
RUO – Research Use Only