missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic acid, 99%, for analysis, Thermo Scientific Chemicals

Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated

$38.02 - $506.77

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC270480250
SDS View Documents
Thermo Scientific Chemicals
270480250
25 mL Plastic Bottle
Each for $38.02
 
AC270480010
SDS Accessories View Documents Promotion Details
Thermo Scientific Chemicals
270480010
1 L Plastic Bottle
Each for $223.89
 
AC270480025
SDS View Documents
Thermo Scientific Chemicals
270480025
2.5 L Plastic Bottle
Each for $506.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Chemical Identifiers

64-18-6
46.025
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
CH2O2
MFCD00003297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O

Specifications

8.0°C
101.0°C
CH2O2
MFCD00003297
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.02
Analytical
25 mL
Authentic
Liquid
1.22
Formic acid
64-18-6
99%
HCO2H
02, 8
15, 4269
Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine
C(=O)O
46.025
CHEBI:30751
99%
1.2200g/mL
50°C
Plastic Bottle
1.3700 to 1.3720
44mbar at 20°C
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only