Learn More
Formic acid, ACS, 96+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03661736
Description
Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Formic Acid | |
7°C to 9°C | |
100°C to 101°C | |
CH2O2 | |
MFCD00003297 | |
1209246 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.03 | |
ACS | |
500 g | |
Liquid | |
Hygroscopic |
For analysis ACS | |
64-18-6 | |
>96+% | |
HCOOH | |
UN1779 | |
14,4241 | |
Miscible with water. Partially miscible in toluene,xylene and benzene. | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥96% | |
1.220 g/mL | |
50°C (156°F) | |
1.371 |
Chemical Identifiers
64-18-6 | |
46.025 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
MFCD00003297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 | |
C(=O)O |
Safety and Handling
GHS H Statement
H331-H314-H318-H226-H302
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P331-P363-P372-P374-P380-P501c
H226-H302-H314-H331
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: FORMIC ACID
EINECSNumber : 200-579-1
RTECSNumber : LQ4900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only